siRNA Details
Gene Symbol: RPS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011.3 | NP_001002.1 | 4, 5 | 398 | ||
GenBank mRNA | AK311794.1 | - | 4, 5 | 336 | ||
BC002866.2 | - | 4, 5 | 354 | 3940495 | ||
BC061901.1 | - | 4, 5 | 391 | 4442840 | ||
BC071919.1 | - | 1 | 318 | 5811776 | ||
BG615667.1 | - | 4, 5 | 399 | |||
M77233.1 | - | 3, 4 | 287 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
