siRNA Details
Gene Symbol: KRTAP2-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033184.3 | NP_149440.1 | 1 | 296 | ||
GenBank mRNA | AJ406930.1 | - | 1 | 296 | ||
BC101147.2 | - | 1 | 292 | 40015180 | ||
BC101148.2 | - | 1 | 292 | 40015187 | ||
BC101149.2 | - | 1 | 292 | 40015188 |
Gene Symbol: KRTAP2-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123387.1 | NP_001116859.1 | 1 | 311 | ||
GenBank mRNA | AJ296345.1 | - | 1 | 167 |
Gene Symbol: KRTAP2-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033032.2 | NP_149021.2 | 1 | 262 | ||
GenBank mRNA | AJ302536.1 | - | 134 |
Gene Symbol: KRTAP2-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165252.1 | NP_001158724.1 | 1 | 304 | ||
GenBank mRNA | BC063625.1 | - | 1 | 304 | 3907481 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
