siRNA Details
Gene Symbol: PGM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021965.3 | NP_068800.2 | 2 | 557 | ||
XM_011518782.2 | XP_011517084.1 | 2 | 847 | |||
XM_011518783.2 | XP_011517085.1 | 2 | 847 | |||
XM_017014808.1 | XP_016870297.1 | 2 | 449 | |||
GenBank mRNA | AK297108.1 | - | 2 | 379 | ||
BC033073.1 | - | 2 | 557 | 4123706 |
Gene Symbol: PGM5P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002836.2 | 2 | 505 | |||
GenBank mRNA | BC025351.1 | - | 2 | 497 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
