siRNA Details
Gene Symbol: SNRPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304464.1 | NP_001291393.1 | 4 | 283 | ||
NM_003094.3 | NP_003085.1 | 4 | 239 | |||
NR_130746.1 | 3 | 242 | ||||
GenBank mRNA | AA344740.1 | - | 3 | 189 | ||
AI263891.1 | - | 3 | 218 | |||
AK307255.1 | - | 4 | 269 | |||
AK312130.1 | - | 4 | 216 | |||
BC002639.2 | - | 4 | 208 | 3609230 | ||
BC072433.1 | - | 4 | 187 | |||
BC090951.1 | - | 4 | 212 | 5204266 | ||
CN272809.1 | - | 3 | 167 | |||
M15919.1 | - | 4 | 132 | |||
M37716.1 | - | 4 | 209 | |||
X12466.1 | - | 4 | 209 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
