siRNA Details
Gene Symbol: GOLGA6L5P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003246.4 | 6 | 1159 | |||
GenBank mRNA | AK092490.1 | - | 3 | 772 | ||
AK097432.1 | - | 8 | 1182 | |||
AK130562.1 | - | 5 | 911 |
Gene Symbol: GOLGA6L10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164465.3 | NP_001157937.2 | 6 | 1113 | ||
NM_001322400.1 | NP_001309329.1 | 9 | 1155 | |||
GenBank mRNA | AK302092.1 | - | 6 | 1075 | ||
CR936602.1 | - | 7, 8 | 1782 |
Gene Symbol: GOLGA6L17P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_111962.1 | 6 | 1077 | |||
GenBank mRNA | AK301885.1 | - | 1203 |
Gene Symbol: GOLGA6L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310153.1 | NP_001297082.1 | 6 | 1096 | ||
GenBank mRNA | AK301968.1 | - | 10 | 1000 |
Gene Symbol: GOLGA6L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267536.2 | NP_001254465.2 | 6 | 1054 | ||
XM_017022481.1 | XP_016877970.1 | 7 | 1054 | |||
XM_017022482.1 | XP_016877971.1 | 7 | 1054 |
Gene Symbol: GOLGA6L9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291420.1 | NP_001278349.1 | 5 | 729 | ||
NM_198181.3 | NP_937824.3 | 6 | 861 | |||
XM_011521601.2 | XP_011519903.1 | 5 | 1412 | |||
GenBank mRNA | BC041854.1 | - | 5 | 729 | 5270411 | |
BY996849.1 | - | 3 | 604 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
