siRNA Details
Gene Symbol: KRTAP6-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181604.1 | NP_853635.1 | 1 | 19 | ||
GenBank mRNA | AB096952.1 | - | 1 | 45 | ||
BC093915.1 | - | 1 | 47 | |||
BC093917.1 | - | 1 | 47 |
Gene Symbol: KRTAP6-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181602.1 | NP_853633.1 | 1 | 19 | ||
GenBank mRNA | AB096954.1 | - | 1 | 45 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
