siRNA Details
Gene Symbol: KRTAP19-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181609.3 | NP_853640.1 | 1 | 32 | ||
GenBank mRNA | AB096944.1 | - | 1 | 60 | ||
AF139541.1 | - | 1 | 32 | |||
BC126280.1 | - | 1 | 76 | |||
BC126282.1 | - | 1 | 76 |
Gene Symbol: KRTAP19-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099219.1 | NP_001092689.1 | 1 | 65 | ||
GenBank mRNA | AB096964.1 | - | 1 | 65 |
Gene Symbol: KRTAP6-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181602.1 | NP_853633.1 | 1 | 68 | ||
GenBank mRNA | AB096954.1 | - | 1 | 94 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
