siRNA Details
Gene Symbol: KRTAP6-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181605.3 | NP_853636.3 | 1 | 192 | ||
GenBank mRNA | AB096951.1 | - | 1 | 192 | ||
BC101121.2 | - | 1 | 166 | |||
BC101122.1 | - | 1 | 166 | |||
BC118547.1 | - | 1 | 166 | |||
BC118624.1 | - | 1 | 166 |
Gene Symbol: KRTAP20-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181616.1 | NP_853647.1 | 1 | 108 | ||
GenBank mRNA | AB096957.1 | - | 1 | 136 | ||
BC111084.1 | - | 1 | 108 | 40013739 |
Gene Symbol: KRTAP6-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181602.1 | NP_853633.1 | 1 | 93 | ||
GenBank mRNA | AB096954.1 | - | 1 | 119 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
