siRNA Details
Gene Symbol: ZNF658 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317916.1 | NP_001304845.1 | 3 | 357 | ||
NM_033160.6 | NP_149350.3 | 3 | 200 | |||
NR_134255.1 | 2 | 245 | ||||
XM_005272515.4 | XP_005272572.1 | 3 | 307 | |||
XM_011545679.2 | XP_011543981.1 | 3 | 131 | |||
XM_017014614.1 | XP_016870103.1 | 2 | 2493 | |||
GenBank mRNA | AK057375.1 | - | 3 | 147 | ||
AK292150.1 | - | 3 | 339 | |||
BC031626.1 | - | 3 | 281 | 5172270 | ||
DA103495.1 | - | 3 | 201 | |||
DA120232.1 | - | 3 | 139 | |||
DA404383.1 | - | 3 | 197 |
Gene Symbol: ZNF658B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DA115965.1 | - | 3 | 192 | ||
DR001461.1 | - | 3 | 186 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
