siRNA Details
Gene Symbol: ZNF658 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317916.1 | NP_001304845.1 | 3, 4 | 460 | ||
NM_033160.6 | NP_149350.3 | 3, 4 | 303 | |||
NR_134255.1 | 2, 3 | 348 | ||||
XM_005272515.4 | XP_005272572.1 | 3, 4 | 410 | |||
XM_011545679.2 | XP_011543981.1 | 3, 4 | 234 | |||
XM_017014614.1 | XP_016870103.1 | 2, 3 | 2596 | |||
GenBank mRNA | AK057375.1 | - | 3, 4 | 250 | ||
AK292150.1 | - | 3, 4 | 442 | |||
BC031626.1 | - | 3, 4 | 384 | 5172270 | ||
DA103495.1 | - | 3, 4 | 304 | |||
DA120232.1 | - | 3, 4 | 242 | |||
DA404383.1 | - | 3, 4 | 300 |
Gene Symbol: ZNF658B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003528.3 | 3, 4 | 299 | |||
GenBank mRNA | DA115965.1 | - | 3, 4 | 295 | ||
DR001461.1 | - | 3, 4 | 289 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
