siRNA Details
Gene Symbol: BMPR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004329.2 | NP_004320.2 | 8 | 1168 | ||
XM_011540103.2 | XP_011538405.1 | 8 | 1065 | |||
XM_011540104.2 | XP_011538406.1 | 9 | 1232 | |||
GenBank mRNA | AB451405.1 | - | 6 | 620 | ||
AK291764.1 | - | 8 | 1037 | |||
BC028383.1 | - | 8 | 1162 | 4824378 | ||
Z22535.1 | - | 8 | 929 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
