siRNA Details
Gene Symbol: LINC00341 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026779.1 | 1 | 575 | |||
GenBank mRNA | AK024929.1 | - | 1 | 173 | ||
BC008299.1 | - | 1 | 575 | |||
CR457337.1 | - | 1 | 124 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
