siRNA Details
Gene Symbol: RPL36AL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001.4 | NP_000992.1 | 2 | 445 | ||
GenBank mRNA | AB063609.1 | - | 2 | 391 | ||
AK311959.1 | - | 2 | 390 | |||
BC000741.2 | - | 2 | 378 | 3507009 | ||
BC003145.1 | - | 2 | 371 | 2957923 | ||
BC070207.1 | - | 2 | 380 | 4271403 | ||
BC107716.1 | - | 2 | 405 | 6727975 | ||
CR542166.1 | - | 1 | 285 | |||
M15661.1 | - | 1 | 315 |
Gene Symbol: RPL36A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021029.5 | NP_066357.2 | 4 | 439 | ||
GenBank mRNA | AY927233.1 | - | 1 | 302 | ||
BC001781.1 | - | 1 | 325 | 3353669 | ||
BC031015.1 | - | 1 | 317 | 4659712 | ||
BC062219.1 | - | 4 | 350 | 6726961 | ||
BC070204.1 | - | 1 | 314 | 4774945 | ||
BF211310.1 | - | 4 | 370 | |||
BG504049.1 | - | 4 | 318 | |||
BP245950.1 | - | 4 | 439 | |||
CR542198.1 | - | 1 | 285 |
Gene Symbol: RPL36A-HNRNPH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199973.1 | NP_001186902.1 | 4 | 439 | ||
GenBank mRNA | DA998453.1 | - | 4 | 319 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
