siRNA Details
Gene Symbol: DTWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144955.1 | NP_001138427.1 | 2 | 220 | ||
NM_020234.5 | NP_064619.2 | 3 | 348 | |||
XM_011521815.1 | XP_011520117.1 | 2 | 269 | |||
XM_017022419.1 | XP_016877908.1 | 2 | 267 | |||
XM_017022420.1 | XP_016877909.1 | 3 | 1129 | |||
XM_017022421.1 | XP_016877910.1 | 3 | 360 | |||
XM_017022422.1 | XP_016877911.1 | 2 | 1001 | |||
XM_017022423.1 | XP_016877912.1 | 2 | 269 | |||
XM_017022424.1 | XP_016877913.1 | 3 | 340 | |||
XM_017022425.1 | XP_016877914.1 | 3 | 340 | |||
XM_017022426.1 | XP_016877915.1 | 2 | 1001 | |||
XM_017022427.1 | XP_016877916.1 | 2 | 227 | |||
XM_017022428.1 | XP_016877917.1 | 2 | 232 | |||
XM_017022429.1 | XP_016877918.1 | 2 | 1001 | |||
XM_017022430.1 | XP_016877919.1 | 2 | 204 | |||
GenBank mRNA | AF168717.1 | - | 2 | 197 | ||
AK290184.1 | - | 2 | 189 | |||
BC018028.1 | - | 2 | 209 | |||
BC032535.2 | - | 3 | 288 | 5527713 | ||
BC093073.1 | - | 2 | 175 | 30326731 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
