siRNA Details
Gene Symbol: GH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000515.4 | NP_000506.2 | 5 | 581 | ||
NM_022559.3 | NP_072053.1 | 5 | 536 | |||
NM_022560.3 | NP_072054.1 | 4 | 461 | |||
GenBank mRNA | A00469.1 | - | 5 | 544 | ||
AF110644.1 | - | 5 | 377 | |||
AF185611.1 | - | 6 | 409 | |||
AV753220.1 | - | 3 | 401 | |||
AY613431.1 | - | 5 | 567 | |||
AY613432.1 | - | 5 | 567 | |||
BC062475.1 | - | 5 | 501 | 30393581 | ||
BC075012.2 | - | 5 | 532 | 30915404 | ||
BC075013.2 | - | 5 | 532 | 30915617 | ||
BC090045.1 | - | 5 | 548 | 30394115 | ||
CD108592.1 | - | 4 | 427 | |||
V00519.1 | - | 5 | 545 |
Gene Symbol: CSH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317.5 | NP_001308.1 | 5 | 655 | ||
GenBank mRNA | AK291587.1 | - | 5 | 574 | ||
AU134786.2 | - | 5 | 655 | |||
BC002717.2 | - | 5 | 548 | 3631916 | ||
BC005921.1 | - | 5 | 567 | 4043768 | ||
BC020756.1 | - | 5 | 623 | 4769769 | ||
BC057768.1 | - | 5 | 567 | 30337725 | ||
BC062775.1 | - | 5 | 571 | 4604848 | ||
BG436049.1 | - | 5 | 568 | |||
BT006926.1 | - | 4 | 505 | |||
J00118.1 | - | 5 | 534 | |||
M25118.1 | - | 1 | 28 | |||
V00593.1 | - | 4 | 357 |
Gene Symbol: CSH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020991.3 | NP_066271.1 | 5 | 620 | ||
NM_022644.3 | NP_072170.1 | 4 | 873 | |||
NM_022645.2 | NP_072171.1 | 3 | 335 | |||
GenBank mRNA | BC022044.1 | - | 5 | 571 | 4769046 | |
BC035965.1 | - | 5 | 567 | 4731880 | ||
BC119748.2 | - | 5 | 564 | 40119242 | ||
V00573.1 | - | 5 | 529 |
Gene Symbol: CSHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318.3 | NP_001309.3 | 4 | 476 | ||
NM_001321067.1 | NP_001307996.1 | 4 | 472 | |||
NM_001321068.1 | NP_001307997.1 | 5 | 466 | |||
NM_001321069.1 | NP_001307998.1 | 5 | 535 | |||
NM_022579.2 | NP_072101.1 | 5 | 652 | |||
NM_022580.2 | NP_072102.1 | 4 | 403 | |||
NM_022581.2 | NP_072103.1 | 5 | 583 | |||
XM_011524344.1 | XP_011522646.1 | 4 | 835 | |||
XM_011524346.2 | XP_011522648.1 | 3 | 677 | |||
GenBank mRNA | AX772828.1 | - | 5 | 480 | ||
BC029365.1 | - | 4 | 406 | 4619624 | ||
BC119747.2 | - | 4 | 403 | 40119241 | ||
BX382469.2 | - | 3 | 729 | |||
BX426214.2 | - | 5 | 448 | |||
DR004701.1 | - | 5 | 376 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
