siRNA Details
Gene Symbol: CSH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317.5 | NP_001308.1 | 2 | 310 | ||
GenBank mRNA | AK291587.1 | - | 2 | 229 | ||
AU134786.2 | - | 2 | 310 | |||
BC002717.2 | - | 2 | 203 | 3631916 | ||
BC005921.1 | - | 2 | 222 | 4043768 | ||
BC020756.1 | - | 2 | 278 | 4769769 | ||
BC057768.1 | - | 2 | 222 | 30337725 | ||
BC062775.1 | - | 2 | 226 | 4604848 | ||
BG436049.1 | - | 2 | 223 | |||
BT006926.1 | - | 1 | 160 | |||
J00118.1 | - | 2 | 189 | |||
V00593.1 | - | 1 | 12 |
Gene Symbol: CSH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020991.3 | NP_066271.1 | 2 | 275 | ||
NM_022644.3 | NP_072170.1 | 2 | 275 | |||
NM_022645.2 | NP_072171.1 | 2 | 275 | |||
GenBank mRNA | BC022044.1 | - | 2 | 226 | 4769046 | |
BC035965.1 | - | 2 | 222 | 4731880 | ||
BC119748.2 | - | 2 | 219 | 40119242 | ||
V00573.1 | - | 2 | 184 |
Gene Symbol: CSHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022579.2 | NP_072101.1 | 2 | 292 | ||
NM_022581.2 | NP_072103.1 | 2 | 292 | |||
XM_011524344.1 | XP_011522646.1 | 2 | 222 | |||
GenBank mRNA | AK300190.1 | - | 2 | 222 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
