siRNA Details
Gene Symbol: UPK3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030570.3 | NP_085047.1 | 5 | 1567 | ||
NM_182683.2 | NP_872624.1 | 6 | 1402 | |||
NM_182684.2 | NP_872625.1 | 7 | 1482 | |||
GenBank mRNA | AK056760.1 | - | 6 | 1210 | ||
AK290096.1 | - | 5 | 1470 | |||
AY233462.1 | - | 7 | 1315 | |||
BC004304.1 | - | 5 | 1391 | 3625247 |
Gene Symbol: ADCY10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK310531.1 | - | 9 | 1086 |
Gene Symbol: ADIPOQ-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046662.1 | 3 | 3392 | |||
GenBank mRNA | BC036509.1 | - | 4 | 3528 |
Gene Symbol: DTX2P1-UPK3BP1-PMS2P11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023383.1 | 10 | 1575 |
Gene Symbol: LOC101929881 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR749549.1 | - | 1 | 4105 |
Gene Symbol: LOC441259 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC075816.1 | - | 6 | 617 |
Gene Symbol: PMS2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003613.1 | 6 | 1053 | |||
GenBank mRNA | AB017004.1 | - | 6 | 1053 | ||
D38435.1 | - | 6 | 656 | |||
D38499.1 | - | 6 | 656 |
Gene Symbol: PMS2P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028059.1 | 9 | 1447 | |||
GenBank mRNA | D38437.1 | - | 5 | 595 | ||
D38498.1 | - | 5 | 595 |
Gene Symbol: PMS2P4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_022007.2 | 5 | 545 | |||
NR_046297.1 | 5 | 545 | ||||
NR_046299.1 | 5 | 545 | ||||
GenBank mRNA | BC029368.1 | - | 5 | 522 | ||
CB141754.1 | - | 5 | 510 | |||
D38438.1 | - | 5 | 514 | |||
D38500.1 | - | 5 | 514 |
Gene Symbol: PMS2P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027775.2 | 6 | 662 | |||
NR_027776.2 | 5 | 559 | ||||
GenBank mRNA | BF984173.1 | - | 5 | 558 | ||
D38436.1 | - | 6 | 643 | |||
D38439.1 | - | 4 | 465 | |||
D38501.1 | - | 4 | 465 | |||
D38502.1 | - | 6 | 643 |
Gene Symbol: PMS2P6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB017005.1 | - | 2 | 212 |
Gene Symbol: PMS2P9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028058.1 | 4 | 950 | |||
GenBank mRNA | BM845848.1 | - | 5 | 491 |
Gene Symbol: UPP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135098.1 | NP_001128570.1 | 2 | 147 | ||
GenBank mRNA | BC033529.2 | - | 2 | 147 | 4818300 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
