siRNA Details
Gene Symbol: PSME2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002818.2 | NP_002809.2 | 10, 11 | 702 | ||
XM_006720213.1 | XP_006720276.1 | 10, 11 | 1496 | |||
GenBank mRNA | AK026580.1 | - | 10, 11 | 723 | ||
AK225876.1 | - | 10, 11 | 665 | |||
AY771595.1 | - | 10, 11 | 651 | |||
BC004368.1 | - | 10, 11 | 652 | 3640584 | ||
BC019885.1 | - | 10, 11 | 652 | 4994572 | ||
BC072025.1 | - | 10, 11 | 651 | 6270762 | ||
BX161498.1 | - | 10, 11 | 1277 | |||
CR541657.1 | - | 10, 11 | 637 | |||
CR541743.1 | - | 10, 11 | 637 | |||
D45248.1 | - | 10, 11 | 702 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
