siRNA Details
Gene Symbol: POM121L8P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024583.1 | 1 | 1552 | |||
GenBank mRNA | AK302811.1 | - | 1 | 1555 | ||
AK308294.1 | - | 1 | 502 |
Gene Symbol: LOC727983 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC112340.1 | - | 1 | 1217 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
