siRNA Details
Gene Symbol: FAM156A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242489.1 | NP_001229418.1 | 3 | 1021 | ||
NM_001242490.1 | NP_001229419.1 | 4 | 1005 | |||
NM_001242491.1 | NP_001229420.1 | 3 | 968 | |||
NM_001242492.1 | NP_001229421.1 | 5 | 854 | |||
NM_001242493.1 | NP_001229422.1 | 5 | 747 | |||
NM_001242494.1 | NP_001229423.1 | 4 | 705 | |||
NM_001242495.1 | NP_001229424.1 | 3 | 612 | |||
NM_001242496.1 | NP_001229425.1 | 4 | 596 | |||
NM_001242497.1 | NP_001229426.1 | 3 | 503 | |||
NM_014138.4 | NP_054857.2 | 5 | 1156 | |||
XM_006724586.3 | XP_006724649.1 | 4 | 2253 | |||
XM_006724587.3 | XP_006724650.1 | 4 | 1844 | |||
XM_011530781.1 | XP_011529083.1 | 4 | 1061 | |||
XM_011530782.1 | XP_011529084.1 | 4 | 1330 | |||
XM_011530783.1 | XP_011529085.1 | 4 | 788 | |||
XM_011530784.2 | XP_011529086.1 | 3 | 557 | |||
XM_017029457.1 | XP_016884946.1 | 5 | 3012 | |||
XM_017029458.1 | XP_016884947.1 | 5 | 2603 | |||
GenBank mRNA | AF090943.1 | - | 1 | 196 | ||
AF370413.1 | - | 3 | 1170 | |||
AK057692.1 | - | 3 | 951 | |||
AK092365.1 | - | 4 | 1300 | |||
AK096162.1 | - | 3 | 1456 | |||
AL834135.1 | - | 3 | 620 | |||
BC000867.1 | - | 1 | 171 | 3460178 | ||
BC067819.1 | - | 3 | 604 | 5313567 | ||
CR749619.1 | - | 2 | 461 | |||
DA368012.1 | - | 3 | 486 |
Gene Symbol: FAM156B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099684.2 | NP_001093154.1 | 3 | 944 | ||
NM_001321178.1 | NP_001308107.1 | 3 | 2710 | |||
NM_001321179.1 | NP_001308108.1 | 4 | 1969 | |||
NM_001321180.1 | NP_001308109.1 | 4 | 1107 | |||
NM_001321181.1 | NP_001308110.1 | 4 | 1079 | |||
NM_001321182.1 | NP_001308111.1 | 3 | 968 | |||
NM_001321183.1 | NP_001308112.1 | 4 | 788 | |||
NM_001321186.1 | NP_001308115.1 | 4 | 670 | |||
NM_001321187.1 | NP_001308116.1 | 4 | 652 | |||
NM_001321188.1 | NP_001308117.1 | 4 | 596 | |||
XM_011530817.1 | XP_011529119.1 | 4 | 1061 | |||
XM_011530821.2 | XP_011529123.1 | 3 | 569 | |||
GenBank mRNA | AK091598.1 | - | 4 | 1865 | ||
BX649013.1 | - | 3 | 2673 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
