siRNA Details
Gene Symbol: FAM156B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099684.2 | NP_001093154.1 | 3 | 1846 | ||
NM_001321178.1 | NP_001308107.1 | 3 | 3612 | |||
NM_001321179.1 | NP_001308108.1 | 4 | 2871 | |||
NM_001321180.1 | NP_001308109.1 | 4 | 2009 | |||
NM_001321181.1 | NP_001308110.1 | 4 | 1981 | |||
NM_001321182.1 | NP_001308111.1 | 3 | 1870 | |||
NM_001321183.1 | NP_001308112.1 | 4 | 1690 | |||
NM_001321186.1 | NP_001308115.1 | 4 | 1572 | |||
NM_001321187.1 | NP_001308116.1 | 4 | 1554 | |||
NM_001321188.1 | NP_001308117.1 | 4 | 1498 | |||
XM_011530817.1 | XP_011529119.1 | 4 | 1963 | |||
XM_011530821.2 | XP_011529123.1 | 3 | 1471 | |||
GenBank mRNA | AK091598.1 | - | 4 | 2767 | ||
BX649013.1 | - | 3 | 3575 |
Gene Symbol: FAM156A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242489.1 | NP_001229418.1 | 3 | 1923 | ||
NM_001242490.1 | NP_001229419.1 | 4 | 1907 | |||
NM_001242491.1 | NP_001229420.1 | 3 | 1870 | |||
NM_001242492.1 | NP_001229421.1 | 5 | 1756 | |||
NM_001242493.1 | NP_001229422.1 | 5 | 1649 | |||
NM_001242494.1 | NP_001229423.1 | 4 | 1607 | |||
NM_001242495.1 | NP_001229424.1 | 3 | 1514 | |||
NM_001242496.1 | NP_001229425.1 | 4 | 1498 | |||
NM_001242497.1 | NP_001229426.1 | 3 | 1405 | |||
NM_014138.4 | NP_054857.2 | 5 | 2058 | |||
XM_006724586.3 | XP_006724649.1 | 4 | 3155 | |||
XM_006724587.3 | XP_006724650.1 | 4 | 2746 | |||
XM_011530781.1 | XP_011529083.1 | 4 | 1963 | |||
XM_011530782.1 | XP_011529084.1 | 4 | 2232 | |||
XM_011530783.1 | XP_011529085.1 | 4 | 1690 | |||
XM_011530784.2 | XP_011529086.1 | 3 | 1459 | |||
XM_017029457.1 | XP_016884946.1 | 5 | 3914 | |||
XM_017029458.1 | XP_016884947.1 | 5 | 3505 | |||
GenBank mRNA | AF090943.1 | - | 1 | 1098 | ||
AF370413.1 | - | 3 | 2074 | |||
AK054721.1 | - | 3 | 2484 | |||
AK057692.1 | - | 3 | 1853 | |||
AK092365.1 | - | 4 | 2202 | |||
AK096162.1 | - | 3 | 2358 | |||
AL834135.1 | - | 3 | 1522 | |||
BC000867.1 | - | 1 | 1073 | 3460178 | ||
BC067819.1 | - | 3 | 1506 | 5313567 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
