siRNA Details
Gene Symbol: NXF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022053.3 | NP_071336.1 | 5 | 618 | ||
GenBank mRNA | AF285596.1 | - | 2 | 217 | ||
AJ277659.1 | - | 9 | 1104 | |||
AK000423.1 | - | 1 | 14 | |||
AK308255.1 | - | 5 | 557 | |||
BC015020.1 | - | 5 | 538 | 3921074 |
Gene Symbol: NXF2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099686.2 | NP_001093156.1 | 5 | 618 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
