siRNA Details
Gene Symbol: NXF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032946.2 | NP_116564.2 | 5, 6 | 593 | ||
NR_028089.1 | 5, 6 | 593 | ||||
GenBank mRNA | AJ277654.1 | - | 5, 6 | 593 | ||
AJ277655.1 | - | 5, 6 | 593 | |||
AJ277656.1 | - | 5, 6 | 593 | |||
BC131708.1 | - | 4, 5 | 414 | 40121743 |
Gene Symbol: NXF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022053.3 | NP_071336.1 | 7, 8 | 880 | ||
GenBank mRNA | AF285596.1 | - | 4, 5 | 479 | ||
AJ277659.1 | - | 11, 12 | 1366 | |||
AK000423.1 | - | 3, 4 | 276 | |||
AK308255.1 | - | 7, 8 | 819 | |||
BC015020.1 | - | 7, 8 | 800 | 3921074 |
Gene Symbol: NXF2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099686.2 | NP_001093156.1 | 7, 8 | 880 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
