siRNA Details
Gene Symbol: KPNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320611.1 | NP_001307540.1 | 11 | 2263 | ||
NM_002266.3 | NP_002257.1 | 11 | 1905 | |||
GenBank mRNA | AK024833.1 | - | 11 | 1773 | ||
AK058111.1 | - | 11 | 1774 | |||
AK291954.1 | - | 11 | 1772 | |||
BC013257.1 | - | 11 | 1768 | |||
BC034443.1 | - | 11 | 1731 | |||
BC053343.1 | - | 11 | 1761 | 4450470 | ||
BC067848.1 | - | 11 | 2004 | 4825235 | ||
BC146905.1 | - | 11 | 1759 | 9056833 | ||
U09559.1 | - | 11 | 1775 | |||
U28386.1 | - | 11 | 1775 |
Gene Symbol: LOC146880 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026899.1 | 13 | 2661 | |||
GenBank mRNA | AK055342.1 | - | 12 | 2532 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
