siRNA Details
Gene Symbol: KRT18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000224.2 | NP_000215.1 | 2, 3 | 612 | ||
NM_199187.1 | NP_954657.1 | 3, 4 | 566 | |||
GenBank mRNA | AK129587.1 | - | 3, 4 | 488 | ||
AK223093.1 | - | 2, 3 | 554 | |||
AK308506.1 | - | 3, 4 | 551 | |||
AK313988.1 | - | 2, 3 | 555 | |||
AY762101.1 | - | 2, 3 | 501 | |||
BC000180.2 | - | 2, 3 | 538 | 2984862 | ||
BC004253.1 | - | 2, 3 | 523 | 3631540 | ||
BC008636.1 | - | 2, 3 | 536 | 3463780 | ||
BC020982.1 | - | 2, 3 | 546 | 3850724 | ||
BC072017.1 | - | 2, 3 | 542 | 6251574 | ||
BG753529.1 | - | 3, 4 | 566 | |||
BT019412.1 | - | 2, 3 | 497 | |||
CD106591.1 | - | 2, 3 | 612 | |||
X12881.1 | - | 2, 3 | 543 | |||
X12883.1 | - | 2, 3 | 479 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
