siRNA Details
Gene Symbol: NOMO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004067.3 | NP_001004067.1 | 20 | 2424 | ||
XM_005255318.1 | XP_005255375.1 | 20 | 2418 | |||
GenBank mRNA | BC006531.2 | - | 9 | 1045 |
Gene Symbol: NOMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014287.3 | NP_055102.3 | 20 | 2355 | ||
GenBank mRNA | BC065535.1 | - | 20 | 2308 | 6007603 | |
X57398.1 | - | 20 | 2193 |
Gene Symbol: NOMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004060.1 | NP_001004060.1 | 20 | 2361 | ||
NM_173614.2 | NP_775885.1 | 20 | 2361 | |||
GenBank mRNA | AK225803.1 | - | 20 | 2331 | ||
AL832855.1 | - | 20 | 2355 | |||
BC028389.1 | - | 19 | 2221 | 4824724 | ||
BC041131.1 | - | 20 | 2335 | 5757234 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
