siRNA Details
Gene Symbol: SYT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136504.1 | NP_001129976.1 | 2 | 206 | ||
NM_177402.4 | NP_796376.2 | 2 | 196 | |||
XM_011509192.2 | XP_011507494.1 | 2 | 217 | |||
XM_017000309.1 | XP_016855798.1 | 1 | 1963 | |||
XM_017000310.1 | XP_016855799.1 | 1 | 1963 | |||
XM_017000311.1 | XP_016855800.1 | 2 | 206 | |||
XM_017000312.1 | XP_016855801.1 | 2 | 871 | |||
XM_017000313.1 | XP_016855802.1 | 2 | 870 | |||
GenBank mRNA | AK090672.1 | - | 2 | 190 | ||
AK094430.1 | - | 2 | 206 | |||
BC100814.1 | - | 2 | 61 | 40000996 | ||
BC100815.1 | - | 2 | 61 | 40000997 | ||
BC100817.1 | - | 2 | 61 | 40001000 | ||
DA098279.1 | - | 2 | 199 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
