siRNA Details
Gene Symbol: OR56A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005179.2 | NP_001005179.2 | 1 | 473 | ||
GenBank mRNA | BC136986.1 | - | 1 | 519 | 9020978 |
Gene Symbol: OR56A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001917.2 | NP_001001917.2 | 1 | 366 | ||
GenBank mRNA | BC136904.1 | - | 1 | 366 | 9020895 | |
BC136911.1 | - | 1 | 366 | 9020902 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
