siRNA Details
Gene Symbol: CLK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130028.1 | NP_001123500.1 | 13 | 2259 | ||
NM_003992.4 | NP_003983.2 | 13 | 1409 | |||
XM_005254151.3 | XP_005254208.1 | 13 | 2301 | |||
XM_017021906.1 | XP_016877395.1 | 13 | 2352 | |||
XM_017021907.1 | XP_016877396.1 | 13 | 2310 | |||
XM_017021909.1 | XP_016877398.1 | 18 | 6117 | |||
XM_017021910.1 | XP_016877399.1 | 16 | 5805 | |||
GenBank mRNA | AB209535.1 | - | 11 | 3245 | ||
AK026511.1 | - | 6 | 495 | |||
AK091692.1 | - | 13 | 2259 | |||
AK096115.1 | - | 8 | 2104 | |||
AK097982.1 | - | 10 | 2069 | |||
AK122860.1 | - | 9 | 3160 | |||
AK225680.1 | - | 13 | 1379 | |||
AK291983.1 | - | 13 | 1381 | |||
AL833165.1 | - | 10 | 2421 | |||
BC002555.2 | - | 13 | 1381 | 3138580 | ||
BC006103.1 | - | 12 | 1330 | 3957357 | ||
BC019881.1 | - | 13 | 1392 | 5114635 | ||
BT006993.1 | - | 12 | 1354 | |||
L29217.1 | - | 13 | 1410 | |||
L29220.1 | - | 12 | 1313 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
