siRNA Details
Gene Symbol: XAGE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130777.2 | NP_570133.1 | 2 | 273 | ||
XM_011530836.2 | XP_011529138.1 | 2 | 301 | |||
GenBank mRNA | AJ318880.1 | - | 2 | 248 | ||
BC009232.2 | - | 2 | 253 | 3960352 | ||
BC066311.1 | - | 2 | 255 | 30330532 | ||
H53211.1 | - | 2 | 273 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
