siRNA Details
Gene Symbol: XAGE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130777.2 | NP_570133.1 | 2 | 226 | ||
XM_011530836.2 | XP_011529138.1 | 2 | 254 | |||
GenBank mRNA | AJ318880.1 | - | 2 | 201 | ||
BC009232.2 | - | 2 | 206 | 3960352 | ||
BC066311.1 | - | 2 | 208 | 30330532 | ||
H53211.1 | - | 2 | 226 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
