siRNA Details
Gene Symbol: ABHD17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130111.1 | NP_001123583.1 | 2 | 419 | ||
NM_031213.3 | NP_112490.3 | 2 | 419 | |||
XM_011528338.1 | XP_011526640.1 | 1 | 3291 | |||
XM_017027341.1 | XP_016882830.1 | 1 | 3291 | |||
XM_017027342.1 | XP_016882831.1 | 1 | 3291 | |||
GenBank mRNA | AK024419.1 | - | 1 | 1177 | ||
AK074043.1 | - | 1 | 2247 | |||
AK074548.1 | - | 2 | 401 | |||
AK090438.1 | - | 1 | 2050 | |||
AK289533.1 | - | 2 | 389 | |||
BC000158.2 | - | 1 | 157 | 2985707 | ||
BC009256.2 | - | 2 | 369 | 3954724 | ||
BC011667.2 | - | 2 | 277 | 4122276 | ||
BC020512.1 | - | 2 | 333 | 3850505 | ||
BC033749.1 | - | 2 | 376 | 5197958 | ||
BC035961.1 | - | 2 | 395 | 4805422 | ||
BC071644.1 | - | 2 | 355 | 5242996 | ||
BC071876.1 | - | 2 | 342 | 6672380 | ||
BC094816.1 | - | 2 | 340 | 6266705 | ||
DB025570.1 | - | 2 | 419 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
