siRNA Details
Gene Symbol: C16orf52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164579.1 | NP_001158051.1 | 2 | 308 | ||
GenBank mRNA | BC027604.1 | - | 3 | 338 | ||
BC053950.1 | - | 3 | 325 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
