siRNA Details
Gene Symbol: FEM1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018708.2 | NP_061178.1 | 1 | 1401 | ||
GenBank mRNA | AJ311359.1 | - | 1 | 1261 | ||
AK074232.1 | - | 1 | 1359 | |||
AK126532.1 | - | 2 | 1129 | |||
AK315554.1 | - | 1 | 1373 | |||
AY356352.1 | - | 1 | 1355 | |||
BC004988.1 | - | 1 | 1359 | 2957950 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
