siRNA Details
Gene Symbol: GTF2IRD2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY312852.1 | - | 13 | 1592 |
Gene Symbol: GTF2IRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173537.3 | NP_775808.3 | 16 | 2046 | ||
GenBank mRNA | AK095257.1 | - | 13 | 1606 | ||
AK225845.1 | - | 16 | 2003 | |||
AK291431.1 | - | 16 | 2015 | |||
AL833932.1 | - | 1 | 177 | |||
AL834153.1 | - | 3 | 712 | |||
AY260739.1 | - | 16 | 1999 | |||
AY312853.1 | - | 16 | 1999 | |||
AY312854.1 | - | 15 | 1839 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
