siRNA Details
Gene Symbol: IGLL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB027448.1 | - | 1 | 257 | ||
AB064202.1 | - | 320 | ||||
AB064220.1 | - | 1 | 320 | |||
AJ698334.1 | - | 1 | 254 | |||
BC073784.1 | - | 2 | 335 | 4297904 | ||
JF806287.1 | - | 2 | 314 | |||
X57819.1 | - | 2 | 309 |
Gene Symbol: IGLJ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | X57817.1 | - | 2 | 305 |
Gene Symbol: IGLV1-44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC030984.1 | - | 2 | 357 | 4701898 | |
U96394.1 | - | 1 | 254 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
