siRNA Details
Gene Symbol: CYAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC093098.1 | - | 2 | 300 | 30325463 |
Gene Symbol: BMS1P20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027293.2 | 11 | 1911 | |||
GenBank mRNA | AB063986.1 | - | 1 | 195 | ||
AB363340.1 | - | 1 | 195 | |||
AB363397.1 | - | 1 | 195 | |||
AB845401.1 | - | 1 | 195 | |||
AF103652.1 | - | 1 | 175 | |||
AF103699.1 | - | 1 | 163 | |||
AJ233731.1 | - | 1 | 186 | |||
AJ241364.1 | - | 1 | 183 | |||
AJ399840.1 | - | 1 | 195 | |||
AJ399851.1 | - | 1 | 195 | |||
AM181147.1 | - | 1 | 195 | |||
AY043113.1 | - | 1 | 195 | |||
AY043142.1 | - | 1 | 195 | |||
AY942089.1 | - | 1 | 195 | |||
DB472458.1 | - | 2 | 269 | |||
DQ098732.1 | - | 1 | 195 | |||
DQ490727.1 | - | 1 | 195 | |||
DQ840929.1 | - | 1 | 195 | |||
DQ841007.1 | - | 1 | 195 | |||
DQ841091.1 | - | 1 | 195 | |||
KJ134805.1 | - | 1 | 195 | |||
KJ423145.1 | - | 1 | 195 | |||
KP278518.1 | - | 1 | 195 | |||
KT304764.1 | - | 1 | 195 | |||
KU602548.1 | - | 1 | 195 |
Gene Symbol: IGLJ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | X57817.1 | - | 2 | 246 |
Gene Symbol: IGLL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB027448.1 | - | 1 | 198 | ||
AB064202.1 | - | 261 | ||||
AB064220.1 | - | 1 | 261 | |||
AJ698334.1 | - | 1 | 195 | |||
BC012876.2 | - | 2 | 296 | 4149333 | ||
BC073784.1 | - | 2 | 276 | 4297904 | ||
JF806287.1 | - | 2 | 255 | |||
X57819.1 | - | 2 | 250 |
Gene Symbol: IGLV1-44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC030984.1 | - | 2 | 298 | 4701898 | |
U96394.1 | - | 1 | 195 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
