siRNA Details
Gene Symbol: AQP7P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017015366.1 | XP_016870855.1 | 9 | 1302 | ||
XM_017015367.1 | XP_016870856.1 | 8 | 1219 | |||
XM_017015368.1 | XP_016870857.1 | 8 | 1202 | |||
XM_017015369.1 | XP_016870858.1 | 9 | 1285 | |||
XM_017015370.1 | XP_016870859.1 | 5 | 1302 | |||
XM_017015371.1 | XP_016870860.1 | 7 | 1069 | |||
XM_017015372.1 | XP_016870861.1 | 6 | 1381 | |||
XM_017015373.1 | XP_016870862.1 | 6 | 940 | |||
XM_017015374.1 | XP_016870863.1 | 6 | 878 | |||
XM_017015375.1 | XP_016870864.1 | 5 | 794 |
Gene Symbol: AQP7P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002817.2 | 4 | 1195 | |||
GenBank mRNA | AK128231.1 | - | 4 | 1191 | ||
BC070322.1 | - | 4 | 518 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
