siRNA Details
Gene Symbol: AQP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170.2 | NP_001161.1 | 8 | 1353 | ||
NM_001318156.1 | NP_001305085.1 | 7 | 1176 | |||
NM_001318157.1 | NP_001305086.1 | 7 | 2029 | |||
NM_001318158.1 | NP_001305087.1 | 7 | 1711 | |||
NR_134513.1 | 5 | 1468 | ||||
NR_134514.1 | 6 | 1587 | ||||
NR_134515.1 | 7 | 1738 | ||||
XM_005251453.3 | XP_005251510.1 | 9 | 1453 | |||
XM_006716765.3 | XP_006716828.1 | 7 | 1114 | |||
XM_011517866.2 | XP_011516168.1 | 8 | 1263 | |||
XM_011517867.2 | XP_011516169.1 | 6 | 893 | |||
XM_017014700.1 | XP_016870189.1 | 8 | 1554 | |||
XM_017014701.1 | XP_016870190.1 | 7 | 1224 | |||
XM_017014702.1 | XP_016870191.1 | 8 | 1259 | |||
XM_017014703.1 | XP_016870192.1 | 7 | 1012 | |||
XM_017014704.1 | XP_016870193.1 | 6 | 1320 | |||
XM_017014705.1 | XP_016870194.1 | 6 | 990 | |||
XM_017014706.1 | XP_016870195.1 | 8 | 1742 | |||
GenBank mRNA | AB006190.1 | - | 8 | 1190 | ||
AK055744.1 | - | 1 | 1295 | |||
BC062701.1 | - | 7 | 1003 | 30324263 | ||
BC119672.1 | - | 6 | 1419 | |||
BC119673.1 | - | 6 | 1419 | |||
DQ786216.1 | - | 3 | 565 |
Gene Symbol: AQP7P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002817.2 | 4 | 1346 | |||
GenBank mRNA | AK128231.1 | - | 4 | 1342 | ||
BC070322.1 | - | 4 | 669 |
Gene Symbol: AQP7P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017015366.1 | XP_016870855.1 | 9 | 1453 | ||
XM_017015367.1 | XP_016870856.1 | 8 | 1370 | |||
XM_017015368.1 | XP_016870857.1 | 8 | 1353 | |||
XM_017015369.1 | XP_016870858.1 | 9 | 1436 | |||
XM_017015370.1 | XP_016870859.1 | 5 | 1453 | |||
XM_017015371.1 | XP_016870860.1 | 7 | 1220 | |||
XM_017015373.1 | XP_016870862.1 | 6 | 1091 | |||
XM_017015374.1 | XP_016870863.1 | 6 | 1029 | |||
XM_017015375.1 | XP_016870864.1 | 5 | 945 |
Gene Symbol: LOC100509620 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005276965.4 | XP_005277022.1 | 5 | 844 | ||
XM_006712884.3 | XP_006712947.1 | 5 | 1268 | |||
XM_006712887.3 | XP_006712950.1 | 7 | 1221 | |||
XM_017005467.1 | XP_016860956.1 | 4 | 840 | |||
XM_017005468.1 | XP_016860957.1 | 4 | 748 | |||
XM_017005469.1 | XP_016860958.1 | 4 | 737 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
