siRNA Details
Gene Symbol: AQP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170.2 | NP_001161.1 | 3, 4 | 474 | ||
NM_001318157.1 | NP_001305086.1 | 3, 4 | 792 | |||
NM_001318158.1 | NP_001305087.1 | 3, 4 | 474 | |||
XM_005251453.3 | XP_005251510.1 | 4, 5 | 574 | |||
XM_011517866.2 | XP_011516168.1 | 3, 4 | 384 | |||
XM_017014700.1 | XP_016870189.1 | 3, 4 | 675 | |||
GenBank mRNA | AB006190.1 | - | 3, 4 | 311 | ||
AK297860.1 | - | 3, 4 | 379 | |||
AK297885.1 | - | 3, 4 | 608 |
Gene Symbol: AQP7P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017015366.1 | XP_016870855.1 | 4, 5 | 574 | ||
XM_017015367.1 | XP_016870856.1 | 3, 4 | 491 | |||
XM_017015368.1 | XP_016870857.1 | 3, 4 | 474 | |||
XM_017015369.1 | XP_016870858.1 | 4, 5 | 557 |
Gene Symbol: LOC100509620 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006712887.3 | XP_006712950.1 | 2, 3 | 342 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
