siRNA Details
Gene Symbol: CD8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178100.1 | NP_001171571.1 | 2 | 352 | ||
NM_004931.4 | NP_004922.1 | 2 | 352 | |||
NM_172101.3 | NP_742099.1 | 2 | 352 | |||
NM_172102.3 | NP_742100.1 | 2 | 352 | |||
NM_172213.3 | NP_757362.1 | 2 | 352 | |||
XM_011533164.2 | XP_011531466.1 | 2 | 310 | |||
GenBank mRNA | AK303360.1 | - | 2 | 310 | ||
AK303595.1 | - | 1 | 206 | |||
BC100911.1 | - | 2 | 343 | 40003720 | ||
BC100912.2 | - | 2 | 343 | 40003723 | ||
BC100913.1 | - | 2 | 343 | 40003724 | ||
BC100914.1 | - | 2 | 343 | 40003725 | ||
DC405359.1 | - | 2 | 352 | |||
M36712.1 | - | 2 | 338 | |||
X13444.1 | - | 2 | 343 | |||
X13445.1 | - | 2 | 343 | |||
X13446.1 | - | 2 | 343 | |||
X13452.1 | - | 2 | 307 | |||
Y00805.1 | - | 2 | 338 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
