siRNA Details
Gene Symbol: HIST2H3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021059.2 | NP_066403.2 | 1 | 154 | ||
GenBank mRNA | BC015544.1 | - | 1 | 154 | ||
BC074969.2 | - | 1 | 146 | 30915509 | ||
BC130635.1 | - | 1 | 148 | 40146666 | ||
BC130637.1 | - | 1 | 148 | 40146668 |
Gene Symbol: HIST2H3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005464.2 | NP_001005464.1 | 1 | 154 |
Gene Symbol: HIST2H3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123375.2 | NP_001116847.1 | 1 | 118 |
Gene Symbol: HIST3H3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003493.2 | NP_003484.1 | 1 | 118 | ||
GenBank mRNA | AK312217.1 | - | 1 | 151 | ||
BC069079.1 | - | 1 | 118 | 7216893 | ||
BC101837.1 | - | 1 | 166 | 8069343 | ||
BC101839.1 | - | 1 | 166 | 8069345 | ||
CR542013.1 | - | 1 | 118 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
