siRNA Details
Gene Symbol: TMEM236 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098844.2 | NP_001092314.1 | 2, 3 | 362 | ||
XM_011519626.2 | XP_011517928.1 | 2, 3 | 513 | |||
XM_011519627.2 | XP_011517929.1 | 2, 3 | 239 | |||
XM_017016574.1 | XP_016872063.1 | 2, 3 | 1088 | |||
GenBank mRNA | AK300456.1 | - | 1, 2 | 105 | ||
BC157856.1 | - | 2, 3 | 362 | 9057075 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
