siRNA Details
Gene Symbol: TMEM236 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098844.2 | NP_001092314.1 | 3 | 410 | ||
XM_011519626.2 | XP_011517928.1 | 3 | 561 | |||
XM_011519627.2 | XP_011517929.1 | 3 | 287 | |||
XM_011519628.1 | XP_011517930.1 | 2 | 128 | |||
XM_011519629.1 | XP_011517931.1 | 2 | 176 | |||
XM_017016574.1 | XP_016872063.1 | 3 | 1136 | |||
GenBank mRNA | AK300456.1 | - | 2 | 153 | ||
BC157856.1 | - | 3 | 410 | 9057075 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
