siRNA Details
Gene Symbol: TDGF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK313319.1 | - | 2 | 184 | ||
BC022393.1 | - | 2 | 246 | 4615416 | ||
BC067844.1 | - | 2 | 246 | 5286020 | ||
CX755454.1 | - | 2 | 57 | |||
DA941183.1 | - | 2 | 448 |
Gene Symbol: TDGF1P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002718.2 | 1 | 882 | |||
GenBank mRNA | M96956.1 | - | 1 | 873 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
