siRNA Details
Gene Symbol: TDGF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003212.3 | NP_003203.1 | 1, 2 | 425 | ||
GenBank mRNA | AK313319.1 | - | 1, 2 | 161 | ||
AU125451.1 | - | 1 | 360 | |||
BC022393.1 | - | 1, 2 | 223 | 4615416 | ||
BC067844.1 | - | 1, 2 | 223 | 5286020 | ||
DA941183.1 | - | 1, 2 | 425 | |||
X14253.1 | - | 1, 2 | 288 |
Gene Symbol: TDGF1P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002718.2 | 1 | 859 | |||
GenBank mRNA | M96956.1 | - | 1 | 850 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
