siRNA Details
Gene Symbol: TMEM106A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291586.1 | NP_001278515.1 | 9 | 1944 | ||
NM_001291587.1 | NP_001278516.1 | 10 | 1778 | |||
NM_001291588.1 | NP_001278517.1 | 7 | 1613 | |||
NM_145041.3 | NP_659478.1 | 9 | 1922 | |||
XM_006721658.2 | XP_006721721.1 | 8 | 1749 | |||
XM_006721659.3 | XP_006721722.1 | 10 | 1775 | |||
XM_011524260.1 | XP_011522562.1 | 8 | 1735 | |||
XM_017024114.1 | XP_016879603.1 | 9 | 2034 | |||
GenBank mRNA | AK056132.1 | - | 9 | 1874 | ||
BC012139.2 | - | 8 | 1826 | 4562851 | ||
BC146976.1 | - | 8 | 1739 | 9056904 | ||
BC157892.1 | - | 8 | 1738 | 9057097 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
