siRNA Details
Gene Symbol: RPS10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014.4 | NP_001005.1 | 2 | 134 | ||
NM_001203245.2 | NP_001190174.1 | 2 | 286 | |||
NM_001204091.1 | NP_001191020.1 | 2 | 138 | |||
GenBank mRNA | AB209165.1 | - | 1 | 89 | ||
AK311797.1 | - | 2 | 81 | |||
BC001032.2 | - | 2 | 59 | 3139393 | ||
BC001955.1 | - | 2 | 113 | 2905318 | ||
BC005012.1 | - | 2 | 113 | 3637658 | ||
BC070235.1 | - | 2 | 57 | 6453788 | ||
BC071946.1 | - | 2 | 69 | 6305105 | ||
BC073799.1 | - | 2 | 146 | 5517455 | ||
BE397113.1 | - | 2 | 84 | |||
BG532491.1 | - | 2 | 287 | |||
BG940847.1 | - | 2 | 134 | |||
BQ072109.1 | - | 1 | 254 | |||
BU956627.1 | - | 2 | 62 | |||
BY996439.1 | - | 2 | 293 | |||
CD385233.1 | - | 2 | 136 |
Gene Symbol: RPS10-NUDT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202470.2 | NP_001189399.1 | 2 | 134 | ||
GenBank mRNA | BU521288.1 | - | 2 | 57 |
Gene Symbol: RPS10P19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK025353.1 | - | 1 | 2668 |
Gene Symbol: RPS10P7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026667.1 | 1 | 192 | |||
GenBank mRNA | AK055590.1 | - | 1 | 1390 | ||
BC030568.2 | - | 1 | 124 |
Gene Symbol: TFF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BU539534.1 | - | 754 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
