siRNA Details
Gene Symbol: FOXD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207305.4 | NP_997188.2 | 1 | 561 | ||
GenBank mRNA | AY344640.1 | - | 1 | 349 | ||
BC089432.1 | - | 1 | 564 | 30555309 | ||
BC103886.2 | - | 1 | 262 | |||
BC136570.1 | - | 1 | 296 | 9020560 | ||
BC136571.1 | - | 1 | 296 | 9020561 |
Gene Symbol: FOXD4L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012184.4 | NP_036316.1 | 1 | 445 | ||
GenBank mRNA | AF452723.1 | - | 1 | 293 | ||
AK122950.1 | - | 1 | 445 | |||
AK125398.1 | - | 1 | 864 | |||
AY344639.1 | - | 1 | 357 | |||
BC136428.1 | - | 1 | 437 | 9020417 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
