siRNA Details
Gene Symbol: FOXD4L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199135.4 | NP_954586.4 | 1 | 1050 | ||
GenBank mRNA | AY344641.1 | - | 1 | 1050 | ||
AY344642.1 | - | 1 | 1050 |
Gene Symbol: FOXD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207305.4 | NP_997188.2 | 1 | 1251 | ||
GenBank mRNA | AF452724.1 | - | 1 | 231 | ||
AY344640.1 | - | 1 | 1039 | |||
BC089432.1 | - | 1 | 1254 | 30555309 | ||
BC103886.2 | - | 1 | 952 | |||
BC136570.1 | - | 1 | 986 | 9020560 | ||
BC136571.1 | - | 1 | 986 | 9020561 |
Gene Symbol: FOXD4L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012184.4 | NP_036316.1 | 1 | 1136 | ||
GenBank mRNA | AF452723.1 | - | 1 | 984 | ||
AK122950.1 | - | 1 | 1136 | |||
AK125398.1 | - | 1 | 1555 | |||
AY344639.1 | - | 1 | 1048 | |||
BC136428.1 | - | 1 | 1128 | 9020417 |
Gene Symbol: FOXD4L5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126334.1 | NP_001119806.1 | 1 | 1797 |
Gene Symbol: FOXD4L6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085476.1 | NP_001078945.1 | 1 | 1557 | ||
GenBank mRNA | BC103887.2 | - | 1 | 965 | 40004959 | |
BC137365.1 | - | 1 | 1045 | 9021363 | ||
BC137366.1 | - | 1 | 1045 | 9021364 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
